siRNA Details
| Gene Symbol: LINC01467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110074.1 | 2 | 930 | |||
| GenBank mRNA | AK130854.1 | - | 3 | 514 | ||
| BC029835.1 | - | 2 | 930 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB520607.1 | - | 1 | 198 | Chr. 14 - 81605344 - 81607378 [-] on Build GRCh38 | |

LINC01467
Genomic Map