siRNA Details
Gene Symbol: ZNF561-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122038.1 | 3 | 378 | |||
GenBank mRNA | AK097685.1 | - | 3 | 343 | ||
BC143540.1 | - | 2 | 251 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF561-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122038.1 | 3 | 378 | |||
GenBank mRNA | AK097685.1 | - | 3 | 343 | ||
BC143540.1 | - | 2 | 251 |