siRNA Details
Gene Symbol: LOC153684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015447.1 | 1 | 129 | |||
GenBank mRNA | AK123995.1 | - | 1 | 170 | ||
BC101382.1 | - | 1 | 130 | |||
BC101383.2 | - | 1 | 130 | |||
BC101384.1 | - | 1 | 129 | |||
BC101385.2 | - | 1 | 129 |
