siRNA Details
| Gene Symbol: LOC101928254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110182.1 | 5 | 1402 | |||
| NR_110183.1 | 4 | 1215 | ||||
| GenBank mRNA | EF565094.1 | - | 5 | 1410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928254| Gene Symbol: LOC101928254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110182.1 | 5 | 1402 | |||
| NR_110183.1 | 4 | 1215 | ||||
| GenBank mRNA | EF565094.1 | - | 5 | 1410 | ||