siRNA Details
Gene Symbol: LOC100505515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109870.1 | 1 | 288 | |||
GenBank mRNA | HY040996.1 | - | 1 | 289 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU568946.1 | - | 291 | Chr. - - [-] on Build |
