siRNA Details
| Gene Symbol: LOC101927244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125938.1 | 2 | 230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927244| Gene Symbol: LOC101927244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125938.1 | 2 | 230 | |||