siRNA Details
| Gene Symbol: LOC105372397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134907.1 | 3 | 440 | |||
| GenBank mRNA | BM716908.1 | - | 3 | 441 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105372397| Gene Symbol: LOC105372397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134907.1 | 3 | 440 | |||
| GenBank mRNA | BM716908.1 | - | 3 | 441 | ||