siRNA Details
| Gene Symbol: LOC729307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134640.1 | 1 | 335 | |||
| GenBank mRNA | BC031238.1 | - | 1 | 335 | ||
| DB035135.1 | - | 1 | 334 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG773282.1 | - | 1 | 340 | Chr. 4 - 137193756 - 137197497 [+] on Build GRCh38 | |

LOC729307
Genomic Map