siRNA Details
| Gene Symbol: SPACA6P-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108100.1 | 2 | 2695 | |||
| GenBank mRNA | AK125996.1 | - | 2 | 2694 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPACA6P-AS| Gene Symbol: SPACA6P-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108100.1 | 2 | 2695 | |||
| GenBank mRNA | AK125996.1 | - | 2 | 2694 | ||