siRNA Details
| Gene Symbol: LOC729224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135244.1 | 2 | 222 | |||
| GenBank mRNA | BC040292.1 | - | 1 | 1210 | ||
| DB055974.1 | - | 2 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC729224| Gene Symbol: LOC729224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135244.1 | 2 | 222 | |||
| GenBank mRNA | BC040292.1 | - | 1 | 1210 | ||
| DB055974.1 | - | 2 | 174 | |||