siRNA Details
| Gene Symbol: LOC105371046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135176.1 | 3 | 453 | |||
| GenBank mRNA | BX107056.1 | - | 3 | 453 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371046| Gene Symbol: LOC105371046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135176.1 | 3 | 453 | |||
| GenBank mRNA | BX107056.1 | - | 3 | 453 | ||