siRNA Details
| Gene Symbol: TTTY20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001546.1 | 2 | 95 | |||
| GenBank mRNA | AF527832.1 | - | 2 | 95 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF527832.3 | - | 2 | 95 | Chr. Y - 9329880 - 9395788 [-] on Build GRCh38 | |

TTTY20
Genomic Map