siRNA Details
Gene Symbol: TTTY20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001546.1 | 2 | 174 | |||
GenBank mRNA | AF527832.1 | - | 2 | 174 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF527832.3 | - | 2 | 174 | Chr. Y - 9329880 - 9395788 [-] on Build GRCh38 |
