siRNA Details
| Gene Symbol: GPR50-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135300.1 | 2 | 522 | |||
| GenBank mRNA | BX343514.2 | - | 2 | 523 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX324423.2 | - | 2 | 521 | Chr. X - 151175905 - 151177836 [-] on Build GRCh38 | |

GPR50-AS1
Genomic Map