siRNA Details
| Gene Symbol: LOC105378614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135054.1 | 1, 2 | 120 | |||
| GenBank mRNA | AI791463.1 | - | 1, 2 | 150 | ||
| BX106629.1 | - | 1, 2 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378614| Gene Symbol: LOC105378614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135054.1 | 1, 2 | 120 | |||
| GenBank mRNA | AI791463.1 | - | 1, 2 | 150 | ||
| BX106629.1 | - | 1, 2 | 139 | |||