siRNA Details
| Gene Symbol: LINC01360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110676.1 | 3 | 370 | |||
| GenBank mRNA | BC041341.1 | - | 3 | 377 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI464548.1 | - | 3 | 377 | Chr. 1 - 73306168 - 73335599 [+] on Build GRCh38 | |

LINC01360
Genomic Map