siRNA Details
| Gene Symbol: LOC105370424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135255.1 | 2 | 985 | |||
| GenBank mRNA | BC034423.1 | - | 2 | 982 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370424| Gene Symbol: LOC105370424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135255.1 | 2 | 985 | |||
| GenBank mRNA | BC034423.1 | - | 2 | 982 | ||