siRNA Details
| Gene Symbol: LINC00491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103753.1 | 3 | 746 | |||
| NR_103754.1 | 4 | 731 | ||||
| NR_103755.1 | 3 | 668 | ||||
| NR_103756.1 | 2 | 597 | ||||
| GenBank mRNA | AK092078.1 | - | 2 | 888 | ||
| BG777293.1 | - | 2 | 551 | |||
| CN270888.1 | - | 4 | 574 | |||
| DA458115.1 | - | 3 | 565 | |||

LINC00491
Genomic Map