siRNA Details
Gene Symbol: LOC100288866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103773.1 | 1 | 234 | |||
GenBank mRNA | AK303671.1 | - | 1 | 234 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DC411135.1 | - | 1 | 234 | Chr. 17 - 49506310 - 49574064 [-] on Build GRCh38 |
