siRNA Details
| Gene Symbol: LINC01480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110724.1 | 3 | 285 | |||
| GenBank mRNA | AK123333.1 | - | 1 | 1532 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU537857.1 | - | 218 | Chr. - - [-] on Build | ||

LINC01480
Genomic Map