siRNA Details
| Gene Symbol: LOC101929034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104602.1 | 2 | 444 | |||
| GenBank mRNA | BX096909.1 | - | 2 | 395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929034| Gene Symbol: LOC101929034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104602.1 | 2 | 444 | |||
| GenBank mRNA | BX096909.1 | - | 2 | 395 | ||