siRNA Details
| Gene Symbol: ISM1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040043.1 | 1 | 222 | |||
| GenBank mRNA | BC038745.1 | - | 1 | 209 | ||
| DB453950.1 | - | 1 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISM1-AS1| Gene Symbol: ISM1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040043.1 | 1 | 222 | |||
| GenBank mRNA | BC038745.1 | - | 1 | 209 | ||
| DB453950.1 | - | 1 | 222 | |||