siRNA Details
| Gene Symbol: LOC101926892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110653.1 | 3 | 1148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926892| Gene Symbol: LOC101926892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110653.1 | 3 | 1148 | |||