siRNA Details
Gene Symbol: CD81-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108080.1 | 3 | 934 | |||
GenBank mRNA | BC019904.1 | - | 3 | 1224 | ||
BC042822.1 | - | 3 | 930 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CD81-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108080.1 | 3 | 934 | |||
GenBank mRNA | BC019904.1 | - | 3 | 1224 | ||
BC042822.1 | - | 3 | 930 |