siRNA Details
| Gene Symbol: LINC01315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120595.1 | 1 | 59 | |||
| NR_120596.1 | 1 | 59 | ||||
| NR_120597.1 | 1 | 59 | ||||
| GenBank mRNA | BC038245.1 | - | 1 | 25 | ||
| CB050274.1 | - | 1 | 68 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI915409.1 | - | 1 | 25 | Chr. 22 - 42364694 - 42369174 [-] on Build GRCh38 | |

LINC01315
Genomic Map