siRNA Details
| Gene Symbol: SRGAP3-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103443.1 | 3 | 267 | |||
| GenBank mRNA | AK302139.1 | - | 3 | 267 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DC397927.1 | - | 3 | 267 | Chr. 3 - 9216895 - 9218087 [+] on Build GRCh38 | |

SRGAP3-AS3
Genomic Map