siRNA Details
| Gene Symbol: LOC101928820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134633.1 | 4 | 1406 | |||
| NR_134634.1 | 2 | 1232 | ||||
| GenBank mRNA | AK024998.1 | - | 2 | 1232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928820| Gene Symbol: LOC101928820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134633.1 | 4 | 1406 | |||
| NR_134634.1 | 2 | 1232 | ||||
| GenBank mRNA | AK024998.1 | - | 2 | 1232 | ||