siRNA Details
| Gene Symbol: LOC100508046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110505.1 | 2 | 2394 | |||
| GenBank mRNA | AY338953.1 | - | 2 | 2392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100508046| Gene Symbol: LOC100508046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110505.1 | 2 | 2394 | |||
| GenBank mRNA | AY338953.1 | - | 2 | 2392 | ||