siRNA Details
Gene Symbol: LOC101929341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134310.1 | 2 | 646 | |||
GenBank mRNA | BC065517.1 | - | 2 | 646 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM554429.1 | - | 2 | 666 | Chr. 8 - 52194360 - 52199553 [+] on Build GRCh38 |
