siRNA Details
Gene Symbol: FLJ35816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093135.1 | - | 2 | 1467 | ||
BC121811.1 | - | 1 | 1568 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ35816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093135.1 | - | 2 | 1467 | ||
BC121811.1 | - | 1 | 1568 |