siRNA Details
| Gene Symbol: LOC105371795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135646.1 | 1 | 252 | |||
| GenBank mRNA | AK096785.1 | - | 1 | 252 | ||
| DA634757.1 | - | 1 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105371795| Gene Symbol: LOC105371795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135646.1 | 1 | 252 | |||
| GenBank mRNA | AK096785.1 | - | 1 | 252 | ||
| DA634757.1 | - | 1 | 190 | |||