siRNA Details
| Gene Symbol: LOC286083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111948.1 | 1 | 1051 | |||
| GenBank mRNA | BC030648.2 | - | 1 | 1051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC286083| Gene Symbol: LOC286083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111948.1 | 1 | 1051 | |||
| GenBank mRNA | BC030648.2 | - | 1 | 1051 | ||