siRNA Details
Gene Symbol: LINC01365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110660.1 | 5 | 446 | |||
GenBank mRNA | BC038750.1 | - | 5 | 446 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI463491.1 | - | 5 | 451 | Chr. 4 - 119799104 - 119804500 [-] on Build GRCh38 |
