siRNA Details
| Gene Symbol: LINC00869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111950.1 | 7, 8 | 1759 | |||
| GenBank mRNA | AB007962.1 | - | 7, 8 | 1427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00869| Gene Symbol: LINC00869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111950.1 | 7, 8 | 1759 | |||
| GenBank mRNA | AB007962.1 | - | 7, 8 | 1427 | ||