siRNA Details
| Gene Symbol: LINC00970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104091.1 | 5 | 995 | |||
| GenBank mRNA | BC041008.1 | - | 5 | 1000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00970| Gene Symbol: LINC00970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104091.1 | 5 | 995 | |||
| GenBank mRNA | BC041008.1 | - | 5 | 1000 | ||