siRNA Details
| Gene Symbol: LOC101926898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125837.1 | 2, 3 | 252 | |||
| GenBank mRNA | BC042616.1 | - | 2, 3 | 254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926898| Gene Symbol: LOC101926898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125837.1 | 2, 3 | 252 | |||
| GenBank mRNA | BC042616.1 | - | 2, 3 | 254 | ||