siRNA Details
| Gene Symbol: LOC101928453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120638.1 | 1 | 444 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI831798.1 | - | 1 | 444 | Chr. 10 - 14074284 - 14087605 [+] on Build GRCh38 | |

LOC101928453
Genomic Map