siRNA Details
Gene Symbol: LOC101928453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120638.1 | 1 | 142 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI831798.1 | - | 1 | 142 | Chr. 10 - 14074284 - 14087605 [+] on Build GRCh38 |
