siRNA Details
| Gene Symbol: LOC101927472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120622.1 | 1 | 180 | |||
| GenBank mRNA | BM794484.1 | - | 1 | 180 | ||
| BX094877.1 | - | 1 | 157 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927472| Gene Symbol: LOC101927472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120622.1 | 1 | 180 | |||
| GenBank mRNA | BM794484.1 | - | 1 | 180 | ||
| BX094877.1 | - | 1 | 157 | |||