siRNA Details
| Gene Symbol: LOC101927472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120622.1 | 3 | 478 | |||
| GenBank mRNA | BX094877.1 | - | 3 | 455 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927472| Gene Symbol: LOC101927472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120622.1 | 3 | 478 | |||
| GenBank mRNA | BX094877.1 | - | 3 | 455 | ||