siRNA Details
| Gene Symbol: SRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054610.1 | - | 1 | 493 | ||
| AK095112.1 | - | 1 | 2450 | |||
| AK126318.1 | - | 1 | 3820 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SRSF1| Gene Symbol: SRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054610.1 | - | 1 | 493 | ||
| AK095112.1 | - | 1 | 2450 | |||
| AK126318.1 | - | 1 | 3820 | |||