siRNA Details
| Gene Symbol: LINC00317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038872.1 | 3 | 158 | |||
| GenBank mRNA | BF026848.1 | - | 3 | 137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00317| Gene Symbol: LINC00317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038872.1 | 3 | 158 | |||
| GenBank mRNA | BF026848.1 | - | 3 | 137 | ||