siRNA Details
| Gene Symbol: LINC01444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110783.1 | 2 | 671 | |||
| GenBank mRNA | CF995332.1 | - | 2 | 489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01444| Gene Symbol: LINC01444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110783.1 | 2 | 671 | |||
| GenBank mRNA | CF995332.1 | - | 2 | 489 | ||