siRNA Details
| Gene Symbol: TNRC6C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040071.1 | 2 | 2957 | |||
| GenBank mRNA | AK092916.1 | - | 2 | 2030 | ||
| AK094898.1 | - | 2 | 2429 | |||
| AK127699.1 | - | 1 | 2208 | |||
| AY129024.1 | - | 1 | 2717 | |||
| BC041417.2 | - | 2 | 2958 | |||

TNRC6C-AS1
Genomic Map