siRNA Details
| Gene Symbol: TNRC6C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040071.1 | 1 | 1667 | |||
| GenBank mRNA | AK092916.1 | - | 1 | 520 | ||
| AK094898.1 | - | 1 | 1139 | |||
| AK123771.1 | - | 1 | 1845 | |||
| AY129024.1 | - | 1 | 203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNRC6C-AS1| Gene Symbol: TNRC6C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040071.1 | 1 | 1667 | |||
| GenBank mRNA | AK092916.1 | - | 1 | 520 | ||
| AK094898.1 | - | 1 | 1139 | |||
| AK123771.1 | - | 1 | 1845 | |||
| AY129024.1 | - | 1 | 203 | |||