siRNA Details
Gene Symbol: BISPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130765.1 | 5 | 737 | |||
NR_130766.1 | 4 | 629 | ||||
GenBank mRNA | AK311380.1 | - | 4 | 846 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY995843.1 | - | 4 | 260 | Chr. 19 - 17406244 - 17414954 [+] on Build GRCh38 |
