siRNA Details
| Gene Symbol: BISPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130766.1 | 2, 3 | 315 | |||
| GenBank mRNA | AK055623.1 | - | 2, 3 | 258 | ||
| AK311380.1 | - | 2, 3 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BISPR| Gene Symbol: BISPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130766.1 | 2, 3 | 315 | |||
| GenBank mRNA | AK055623.1 | - | 2, 3 | 258 | ||
| AK311380.1 | - | 2, 3 | 532 | |||