siRNA Details
Gene Symbol: FAM99A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026643.1 | 3 | 1028 | |||
GenBank mRNA | AK124823.1 | - | 1 | 1632 | ||
CR627417.1 | - | 3 | 1055 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM99A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026643.1 | 3 | 1028 | |||
GenBank mRNA | AK124823.1 | - | 1 | 1632 | ||
CR627417.1 | - | 3 | 1055 |