siRNA Details
| Gene Symbol: FAM99A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026643.1 | 3 | 1383 | |||
| GenBank mRNA | AK124823.1 | - | 1 | 1987 | ||
| CR627417.1 | - | 3 | 1410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM99A| Gene Symbol: FAM99A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026643.1 | 3 | 1383 | |||
| GenBank mRNA | AK124823.1 | - | 1 | 1987 | ||
| CR627417.1 | - | 3 | 1410 | |||