siRNA Details
| Gene Symbol: LINC01355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110616.1 | 1 | 233 | |||
| GenBank mRNA | AL832122.1 | - | 1 | 1467 | ||
| BX113324.1 | - | 1 | 233 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA252753.1 | - | 1 | 231 | Chr. 1 - 23285040 - 23285517 [-] on Build GRCh38 | |

LINC01355
Genomic Map